siRNA Details
| Gene Symbol: Rhobtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153514.5 | NP_705734.4 | 10 | 2582 | ||
| GenBank mRNA | AF420001.1 | - | 9 | 2073 | ||
| AK038196.1 | - | 10 | 2674 | |||
| AK081100.1 | - | 10 | 2511 | |||
| BC026836.1 | - | 10 | 2563 | 4242008 | ||
| BC056954.1 | - | 10 | 2187 | 5685456 | ||

Rhobtb2
Genomic Map