siRNA Details
| Gene Symbol: Adig | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145635.2 | NP_663610.1 | 2 | 281 | ||
| GenBank mRNA | AK131610.1 | - | 2 | 231 | ||
| AY092026.1 | - | 2 | 287 | |||
| BC054059.1 | - | 2 | 229 | 3373967 | ||
| BE624674.1 | - | 2 | 281 | |||
| DQ223719.1 | - | 2 | 150 | |||

Adig
Genomic Map