siRNA Details
| Gene Symbol: Obox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145709.2 | NP_663755.2 | 2 | 363 | ||
| GenBank mRNA | BC052664.1 | - | 2 | 367 | 30053785 | |
| CJ052752.1 | - | 2 | 365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Obox5| Gene Symbol: Obox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145709.2 | NP_663755.2 | 2 | 363 | ||
| GenBank mRNA | BC052664.1 | - | 2 | 367 | 30053785 | |
| CJ052752.1 | - | 2 | 365 | |||