siRNA Details
| Gene Symbol: Mios | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145374.2 | NP_663349.2 | 3 | 654 | ||
| GenBank mRNA | BC020002.1 | - | 3 | 652 | 3584335 | |
| BU510538.1 | - | 3 | 504 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mios| Gene Symbol: Mios | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145374.2 | NP_663349.2 | 3 | 654 | ||
| GenBank mRNA | BC020002.1 | - | 3 | 652 | 3584335 | |
| BU510538.1 | - | 3 | 504 | |||