siRNA Details
| Gene Symbol: Vmn1r223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083311.1 | NP_001076780.1 | 1 | 50 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r223| Gene Symbol: Vmn1r223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083311.1 | NP_001076780.1 | 1 | 50 | ||