siRNA Details
| Gene Symbol: Vmn1r85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145847.1 | NP_665846.1 | 1 | 446 | ||
| GenBank mRNA | BC110463.1 | - | 1 | 446 | 40040729 | |
| BC110464.1 | - | 1 | 446 | 40040731 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r85| Gene Symbol: Vmn1r85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145847.1 | NP_665846.1 | 1 | 446 | ||
| GenBank mRNA | BC110463.1 | - | 1 | 446 | 40040729 | |
| BC110464.1 | - | 1 | 446 | 40040731 | ||