siRNA Details
| Gene Symbol: Il17f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145856.2 | NP_665855.2 | 2 | 287 | ||
| GenBank mRNA | AB116259.1 | - | 2 | 216 | ||
| AF458064.1 | - | 1 | 192 | |||
| AY380822.1 | - | 2 | 287 | |||
| BC145977.1 | - | 1 | 232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il17f| Gene Symbol: Il17f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145856.2 | NP_665855.2 | 2 | 287 | ||
| GenBank mRNA | AB116259.1 | - | 2 | 216 | ||
| AF458064.1 | - | 1 | 192 | |||
| AY380822.1 | - | 2 | 287 | |||
| BC145977.1 | - | 1 | 232 | |||