siRNA Details
| Gene Symbol: Nod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145857.2 | NP_665856.2 | 4 | 1313 | ||
| GenBank mRNA | AF520774.1 | - | 4 | 1358 | ||
| AK089390.1 | - | 4 | 1277 | |||
| AY160220.1 | - | 3 | 1242 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nod2| Gene Symbol: Nod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145857.2 | NP_665856.2 | 4 | 1313 | ||
| GenBank mRNA | AF520774.1 | - | 4 | 1358 | ||
| AK089390.1 | - | 4 | 1277 | |||
| AY160220.1 | - | 3 | 1242 | |||