siRNA Details
| Gene Symbol: Olfr1116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011734.1 | NP_001011734.1 | 1 | 640 | ||
| Gene Symbol: Olfr1118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207632.2 | NP_997515.2 | 1 | 667 | ||
| GenBank mRNA | BC120792.1 | - | 1 | 667 | 40129715 | |

Olfr1116
Genomic Map