siRNA Details
| Gene Symbol: Olfr1357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011737.2 | NP_001011737.1 | 2 | 460 | ||
| GenBank mRNA | BC147817.1 | - | 1 | 312 | 9087534 | |
| BC147825.1 | - | 1 | 312 | 9087542 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1357| Gene Symbol: Olfr1357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011737.2 | NP_001011737.1 | 2 | 460 | ||
| GenBank mRNA | BC147817.1 | - | 1 | 312 | 9087534 | |
| BC147825.1 | - | 1 | 312 | 9087542 | ||