siRNA Details
| Gene Symbol: Olfr744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011738.1 | NP_001011738.1 | 1 | 907 | ||
| GenBank mRNA | BC138726.1 | - | 1 | 970 | 8861748 | |
| BC138727.1 | - | 1 | 970 | 8861749 | ||
More Information
Set Membership: |
|

Olfr744
Genomic Map