siRNA Details
| Gene Symbol: Olfr1200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005227.2 | NP_001005227.2 | 1 | 589 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1200| Gene Symbol: Olfr1200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005227.2 | NP_001005227.2 | 1 | 589 | ||