siRNA Details
| Gene Symbol: Olfr1386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011741.2 | NP_001011741.1 | 1 | 201 | ||
| GenBank mRNA | BC147372.1 | - | 1 | 201 | 9056436 | |
| BC147373.1 | - | 1 | 201 | 9056431 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1386| Gene Symbol: Olfr1386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011741.2 | NP_001011741.1 | 1 | 201 | ||
| GenBank mRNA | BC147372.1 | - | 1 | 201 | 9056436 | |
| BC147373.1 | - | 1 | 201 | 9056431 | ||