siRNA Details
| Gene Symbol: Olfr324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011743.2 | NP_001011743.2 | 1 | 372 | ||
| GenBank mRNA | BC147575.1 | - | 1 | 372 | 9056565 | |
| BC147579.1 | - | 1 | 372 | 9056569 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr324| Gene Symbol: Olfr324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011743.2 | NP_001011743.2 | 1 | 372 | ||
| GenBank mRNA | BC147575.1 | - | 1 | 372 | 9056565 | |
| BC147579.1 | - | 1 | 372 | 9056569 | ||