siRNA Details
| Gene Symbol: Olfr141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181818.2 | NP_861539.2 | 1 | 569 | ||
| Gene Symbol: Olfr1095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146730.2 | NP_666941.2 | 1 | 569 | ||

Olfr141
Genomic Map