siRNA Details
| Gene Symbol: Olfr314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011760.2 | NP_001011760.1 | 1 | 513 | ||
| GenBank mRNA | BC147421.1 | - | 1 | 512 | 9056475 | |
| BC147422.1 | - | 1 | 513 | 9056493 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr314| Gene Symbol: Olfr314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011760.2 | NP_001011760.1 | 1 | 513 | ||
| GenBank mRNA | BC147421.1 | - | 1 | 512 | 9056475 | |
| BC147422.1 | - | 1 | 513 | 9056493 | ||