siRNA Details
| Gene Symbol: PRKCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316329.1 | NP_001303258.1 | 6 | 933 | ||
| NM_002739.4 | NP_002730.1 | 6 | 933 | |||
| GenBank mRNA | AB209671.1 | - | 6 | 652 | ||
| AF345987.1 | - | 6 | 817 | |||
| AK296412.1 | - | 6 | 941 | |||
| AK296434.1 | - | 6 | 920 | |||
| AK302958.1 | - | 6 | 918 | |||
| AK303741.1 | - | 4 | 340 | |||
| BC047876.1 | - | 6 | 808 | 5764695 | ||
| M13977.1 | - | 6 | 817 | |||
| Z15114.1 | - | 2 | 141 | |||

PRKCG
Genomic Map