siRNA Details
| Gene Symbol: Olfr106-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC145568.1 | - | 1 | 633 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr106-ps| Gene Symbol: Olfr106-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC145568.1 | - | 1 | 633 | ||