siRNA Details
Gene Symbol: Olfr544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC106830.2 | - | 1 | 303 | 40044631 | |
BC126870.1 | - | 1 | 303 | 40044632 | ||
BC126871.1 | - | 1 | 303 | 40044633 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC106830.2 | - | 1 | 303 | 40044631 | |
BC126870.1 | - | 1 | 303 | 40044632 | ||
BC126871.1 | - | 1 | 303 | 40044633 |