siRNA Details
| Gene Symbol: Olfr1287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011773.1 | NP_001011773.1 | 1 | 680 | ||
| Gene Symbol: Olfr1283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207236.1 | NP_997119.1 | 1 | 680 | ||
| GenBank mRNA | BC125536.1 | - | 1 | 723 | 40130051 | |

Olfr1287
Genomic Map