siRNA Details
| Gene Symbol: Olfr1029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011852.2 | NP_001011852.2 | 1 | 832 | ||
| GenBank mRNA | BC150841.1 | - | 1 | 832 | 9087752 | |
| Gene Symbol: Olfr1028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011774.2 | NP_001011774.1 | 1 | 800 | ||

Olfr1029
Genomic Map