siRNA Details
| Gene Symbol: Olfr527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011776.1 | NP_001011776.1 | 1 | 793 | ||
| GenBank mRNA | BC147213.1 | - | 1 | 814 | 9056261 | |
| BC147214.1 | - | 1 | 814 | 9056246 | ||
| Gene Symbol: Olfr531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146953.1 | NP_667164.1 | 1 | 790 | ||
| GenBank mRNA | BC104052.1 | - | 1 | 790 | 40040241 | |
| BC104053.1 | - | 1 | 790 | 40040242 | ||

Olfr527
Genomic Map