siRNA Details
| Gene Symbol: Olfr987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011785.1 | NP_001011785.1 | 1 | 335 | ||
| GenBank mRNA | BC147165.1 | - | 1 | 411 | 9056167 | |
| BC147166.1 | - | 1 | 411 | 9056185 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr987| Gene Symbol: Olfr987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011785.1 | NP_001011785.1 | 1 | 335 | ||
| GenBank mRNA | BC147165.1 | - | 1 | 411 | 9056167 | |
| BC147166.1 | - | 1 | 411 | 9056185 | ||