siRNA Details
| Gene Symbol: Olfr1537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207665.1 | NP_997548.1 | 1 | 802 | ||
| Gene Symbol: Olfr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146829.2 | NP_667040.2 | 2 | 863 | ||

Olfr1537
Genomic Map