siRNA Details
| Gene Symbol: Olfr193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011791.1 | NP_001011791.1 | 1 | 247 | ||
| GenBank mRNA | BC147133.1 | - | 1 | 272 | 9056183 | |
| BC147134.1 | - | 1 | 270 | 9056182 | ||
| Gene Symbol: Olfr191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011807.2 | NP_001011807.2 | 1 | 247 | ||

Olfr193
Genomic Map