siRNA Details
| Gene Symbol: Olfr884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011798.1 | NP_001011798.1 | 1 | 647 | ||
| Gene Symbol: Olfr885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011739.1 | NP_001011739.1 | 1 | 647 | ||

Olfr884
Genomic Map