siRNA Details
| Gene Symbol: Olfr1513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012269.2 | NP_001012269.2 | 1 | 311 | ||
| Gene Symbol: Olfr1512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146432.2 | NP_666643.1 | 1 | 311 | ||
| GenBank mRNA | BC133656.1 | - | 1 | 311 | 40042187 | |

Olfr1513
Genomic Map