siRNA Details
| Gene Symbol: Olfr213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011801.1 | NP_001011801.1 | 1 | 685 | ||
| Gene Symbol: Olfr212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011800.2 | NP_001011800.1 | 2 | 1170 | ||
| GenBank mRNA | AK029829.1 | - | 2 | 1172 | ||
| BC141000.1 | - | 2 | 979 | 9055612 | ||

Olfr213
Genomic Map