siRNA Details
Gene Symbol: Olfr1318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011802.2 | NP_001011802.1 | 1 | 852 | ||
GenBank mRNA | BC147163.1 | - | 1 | 852 | 9056186 | |
BC147164.1 | - | 1 | 852 | 9056168 |
Gene Symbol: Olfr1317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146448.1 | NP_666659.1 | 1 | 797 |
