siRNA Details
| Gene Symbol: Olfr487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011811.1 | NP_001011811.1 | 1 | 526 | ||
| Gene Symbol: Olfr486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146496.1 | NP_666707.1 | 1 | 526 | ||

Olfr487
Genomic Map