siRNA Details
| Gene Symbol: Olfr524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011814.1 | NP_001011814.1 | 1 | 881 | ||
| GenBank mRNA | BC147558.1 | - | 1 | 913 | 9056549 | |
| BC147561.1 | - | 1 | 913 | 9056551 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr524| Gene Symbol: Olfr524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011814.1 | NP_001011814.1 | 1 | 881 | ||
| GenBank mRNA | BC147558.1 | - | 1 | 913 | 9056549 | |
| BC147561.1 | - | 1 | 913 | 9056551 | ||