siRNA Details
| Gene Symbol: Olfr533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011815.1 | NP_001011815.1 | 1 | 891 | ||
| GenBank mRNA | BC147452.1 | - | 1 | 936 | 9056094 | |
| BC147473.1 | - | 1 | 936 | 9056115 | ||
| Gene Symbol: Olfr536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146520.2 | NP_666731.1 | 4 | 1139 | ||
| GenBank mRNA | BC051116.1 | - | 4 | 1139 | 6307473 | |

Olfr533
Genomic Map