siRNA Details
| Gene Symbol: Olfr1181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011816.1 | NP_001011816.1 | 1 | 167 | ||
| Gene Symbol: Olfr1182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011535.2 | NP_001011535.1 | 3 | 427 | ||
| GenBank mRNA | DR065631.1 | - | 3 | 427 | ||

Olfr1181
Genomic Map