siRNA Details
| Gene Symbol: Olfr316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011818.2 | NP_001011818.2 | 1 | 797 | ||
| GenBank mRNA | BC150858.1 | - | 1 | 860 | 9087769 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr316| Gene Symbol: Olfr316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011818.2 | NP_001011818.2 | 1 | 797 | ||
| GenBank mRNA | BC150858.1 | - | 1 | 860 | 9087769 | |