siRNA Details
| Gene Symbol: Olfr312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011819.2 | NP_001011819.1 | 1 | 66 | ||
| GenBank mRNA | BC104268.1 | - | 1 | 66 | 40040766 | |
| BC104273.1 | - | 1 | 66 | 40041031 | ||
| Gene Symbol: Olfr311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146537.2 | NP_666748.2 | 1 | 66 | ||

Olfr312
Genomic Map