siRNA Details
| Gene Symbol: Olfr312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011819.2 | NP_001011819.1 | 1 | 300 | ||
| GenBank mRNA | BC104268.1 | - | 1 | 300 | 40040766 | |
| BC104273.1 | - | 1 | 300 | 40041031 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr312| Gene Symbol: Olfr312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011819.2 | NP_001011819.1 | 1 | 300 | ||
| GenBank mRNA | BC104268.1 | - | 1 | 300 | 40040766 | |
| BC104273.1 | - | 1 | 300 | 40041031 | ||