siRNA Details
| Gene Symbol: Olfr313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146536.2 | NP_666747.2 | 1 | 943 | ||
| GenBank mRNA | BC117002.1 | - | 1 | 943 | 40126321 | |
| BC120742.1 | - | 1 | 943 | 40129665 | ||
| Gene Symbol: Olfr312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011819.2 | NP_001011819.1 | 1 | 861 | ||
| GenBank mRNA | BC104268.1 | - | 1 | 861 | 40040766 | |
| BC104273.1 | - | 1 | 861 | 40041031 | ||

Olfr313
Genomic Map