siRNA Details
| Gene Symbol: Olfr1359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011820.1 | NP_001011820.1 | 1 | 742 | ||
| GenBank mRNA | BC147114.1 | - | 1 | 771 | 9056158 | |
| BC147115.1 | - | 1 | 770 | 9056125 | ||
| Gene Symbol: Olfr1360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146543.2 | NP_666754.2 | 1 | 780 | ||
| GenBank mRNA | BC130243.1 | - | 1 | 746 | ||

Olfr1359
Genomic Map