siRNA Details
| Gene Symbol: Olfr804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011821.1 | NP_001011821.1 | 1 | 825 | ||
| Gene Symbol: Olfr782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011797.1 | NP_001011797.1 | 1 | 825 | ||

Olfr804
Genomic Map