siRNA Details
| Gene Symbol: Olfr834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011823.1 | NP_001011823.1 | 1 | 563 | ||
| Gene Symbol: Olfr832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011824.1 | NP_001011824.1 | 1 | 563 | ||

Olfr834
Genomic Map