siRNA Details
| Gene Symbol: Olfr119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011830.2 | NP_001011830.2 | 2 | 674 | ||
| Gene Symbol: Olfr122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146288.3 | NP_666400.3 | 2 | 671 | ||

Olfr119
Genomic Map