siRNA Details
Gene Symbol: Olfr112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013575.4 | NP_001013593.2 | 3 | 3020 | ||
GenBank mRNA | AK029968.1 | - | 3 | 3018 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013575.4 | NP_001013593.2 | 3 | 3020 | ||
GenBank mRNA | AK029968.1 | - | 3 | 3018 |