siRNA Details
| Gene Symbol: Olfr112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013575.4 | NP_001013593.2 | 3 | 2699 | ||
| GenBank mRNA | AK016338.1 | - | 5 | 1196 | ||
| AK029968.1 | - | 3 | 2697 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr112| Gene Symbol: Olfr112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013575.4 | NP_001013593.2 | 3 | 2699 | ||
| GenBank mRNA | AK016338.1 | - | 5 | 1196 | ||
| AK029968.1 | - | 3 | 2697 | |||