siRNA Details
Gene Symbol: Olfr1500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011831.1 | NP_001011831.1 | 1 | 778 | ||
GenBank mRNA | BC150926.1 | - | 1 | 843 | 9087837 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011831.1 | NP_001011831.1 | 1 | 778 | ||
GenBank mRNA | BC150926.1 | - | 1 | 843 | 9087837 |