siRNA Details
| Gene Symbol: Olfr1129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011836.2 | NP_001011836.2 | 1 | 875 | ||
| Gene Symbol: Olfr1130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146838.2 | NP_667049.2 | 2 | 1055 | ||

Olfr1129
Genomic Map