siRNA Details
| Gene Symbol: Olfr1437 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011839.1 | NP_001011839.1 | 1 | 812 | ||
| Gene Symbol: Olfr1436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146687.2 | NP_666898.2 | 1 | 821 | ||
| GenBank mRNA | BC127991.1 | - | 1 | 825 | 40135681 | |

Olfr1437
Genomic Map