siRNA Details
| Gene Symbol: Olfr591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011847.1 | NP_001011847.1 | 1 | 235 | ||
| Gene Symbol: Olfr593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146380.1 | NP_666492.1 | 1 | 268 | ||

Olfr591
Genomic Map