siRNA Details
| Gene Symbol: Olfr675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011848.1 | NP_001011848.1 | 1 | 746 | ||
| Gene Symbol: Olfr671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011755.1 | NP_001011755.1 | 1 | 746 | ||

Olfr675
Genomic Map