siRNA Details
Gene Symbol: Olfr1028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011774.2 | NP_001011774.1 | 1 | 552 |
Gene Symbol: Olfr1029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011852.2 | NP_001011852.2 | 1 | 584 | ||
GenBank mRNA | BC150841.1 | - | 1 | 584 | 9087752 |
