siRNA Details
| Gene Symbol: Olfr1425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011853.1 | NP_001011853.1 | 1 | 674 | ||
| Gene Symbol: Olfr1424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146681.1 | NP_666892.1 | 1 | 680 | ||

Olfr1425
Genomic Map